missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl phosphate, 99+%

Product Code. 10752705 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
2.5kg
250g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10752705 5 kg Plastic drum 5kg
10051660 250 g Plastic bottle 250g
10561581 1 kg Plastic bottle 1kg
10256333 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10752705 Supplier Thermo Scientific Acros Supplier No. 147670050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115-86-6 | C18H15O4P | 326.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-86-6
Molecular Formula C18H15O4P
Molecular Weight (g/mol) 326.29
MDL Number MFCD00003031
InChI Key XZZNDPSIHUTMOC-UHFFFAOYSA-N
Synonym triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech
PubChem CID 8289
ChEBI CHEBI:35033
IUPAC Name triphenyl phosphate
SMILES O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Acidity 0.05mg KOH/g max.
Melting Point 47°C to 53°C
Color White
Boiling Point 370°C
Flash Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic drum
Linear Formula (C6H5O)3P(O)
Quantity 5 kg
Beilstein 06, 179
Merck Index 15, 9915
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and acetone
Formula Weight 326.28
Percent Purity 99+%
Physical Form Crystalline Flakes
Chemical Name or Material Triphenyl phosphate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.