missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl phosphate, 98%

Product Code. 11377995
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377995 250 g 250g
11387995 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377995 Supplier Thermo Scientific Alfa Aesar Supplier No. L08130.30
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 115-86-6 | C18H15O4P | 326.29 g/mol

Triphenyl phosphate is used as an internal standard in the screening and quantification of agrochemical residues in vegetables and fruits. It is used in plastic processing and molding of plastic flow and performance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyl phosphate is used as an internal standard in the screening and quantification of agrochemical residues in vegetables and fruits. It is used in plastic processing and molding of plastic flow and performance.

Solubility
Reacts with water.

Notes
Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-86-6
Molecular Formula C18H15O4P
Molecular Weight (g/mol) 326.29
MDL Number MFCD00003031
InChI Key XZZNDPSIHUTMOC-UHFFFAOYSA-N
Synonym triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech
PubChem CID 8289
ChEBI CHEBI:35033
IUPAC Name triphenyl phosphate
SMILES O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Melting Point 48°C to 52°C
Boiling Point 247°C to 250°C (11 mmHg)
Flash Point 223°C (433°F)
Odor Odorless
Quantity 250 g
UN Number UN3077
Beilstein 1888236
Merck Index 14,9742
Solubility Information Reacts with water.
Formula Weight 326.29
Percent Purity 98%
Chemical Name or Material Triphenyl phosphate
Show More Show Less
compliance-icons
Product Identifier
  • Triphenyl phosphate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Triphenyl phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.