missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl borate, 97%

Product Code. p-7052759
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15452008

Brand: Thermo Scientific Alfa Aesar H55686.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1095-03-0 | C18H15BO3 | 290.125 g/mol

Triphenyl borate is a useful boronic ester with chemosterilant activity against Cochliomyia hominivorax. It is also used as a competitive inhibitor of urease of Klebsiella aerogenes used to study the mechanisms of interaction with the nickel active site.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyl borate is a useful boronic ester with chemosterilant activity against Cochliomyia hominivorax. It is also used as a competitive inhibitor of urease of Klebsiella aerogenes used to study the mechanisms of interaction with the nickel active site.

Solubility
Soluble in chloroform, methanol.

Notes
Moisture, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic. Store under inert atmosphere.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1095-03-0
Molecular Formula C18H15BO3
Molecular Weight (g/mol) 290.125
MDL Number MFCD00059011
InChI Key MDCWDBMBZLORER-UHFFFAOYSA-N
Synonym triphenoxyborane, phenyl borate, triphenoxyboron, boric acid, triphenyl ester, phenyl borate, pho 3 b, trifenylester kyseliny borite, boric acid h3bo3 , triphenyl ester, triphenyl ester boric acid, trifenylester kyseliny borite czech, boric acid triphenyl ester
PubChem CID 14182
IUPAC Name triphenyl borate
SMILES B(OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3

Especificaciones

Melting Point 98°C to 101°C
Linear Formula (C6H5O)3B
Quantity 5 g
UN Number UN2811
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform,methanol.
Formula Weight 290.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Triphenyl borate
compliance-icons compliance-icons
Nombre del producto
  • Triphenyl borate
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contains : Triphenyl borate

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado