missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Tripeptidyl peptidase II Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29910
This item is not returnable.
View return policy
Description
Tripeptidyl peptidase II Polyclonal antibody specifically detects Tripeptidyl peptidase II in Human samples. It is validated for Immunoprecipitation
Specifications
| Tripeptidyl peptidase II | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| EC 3.4.14.10, FLJ40359, TPP-2, TPP-II, Tripeptidyl aminopeptidase, tripeptidyl peptidase II, tripeptidyl-peptidase 2, Tripeptidyl-peptidase II | |
| Between 375 and 425 | |
| 100 μg | |
| Cell Biology, Immunology | |
| 7174 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction