missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tripeptidyl-Peptidase I/TPP1 Rabbit anti-Human, Mouse, Rat, Clone: S09-2G9, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15105-100UL
This item is not returnable.
View return policy
Description
Tripeptidyl-Peptidase I/TPP1 Monoclonal antibody specifically detects Tripeptidyl-Peptidase I/TPP1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Tripeptidyl-Peptidase I/TPP1 | |
| Monoclonal | |
| Unconjugated | |
| Cell growth-inhibiting gene 1 protein, ceroid-lipofuscinosis, neuronal 2, late infantile (Jansky-Bielschowsky disease), CLN2EC 3.4.14.9, growth-inhibiting protein 1, LPIC, lysosomal pepstatin insensitive protease, Lysosomal pepstatin-insensitive protease, MGC21297, TPP-1, TPP-I, Tripeptidyl aminopeptidase, tripeptidyl peptidase I, tripeptidyl-peptidase 1, Tripeptidyl-peptidase I | |
| Recombinant protein of human Tripeptidyl-Peptidase I/TPP1 (Uniprot#: O14773) | |
| 100 μg | |
| Neuroscience | |
| 1200 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S09-2G9 | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction