missing translation for 'onlineSavingsMsg'
Learn More

Tripalmitin, TRC

Product Code. 30010766
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010766 10 g 10g
30004953 1 g 1g
30010751 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010766 Supplier Toronto Research Chemicals Supplier No. TRCT80813010G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-44-2
Molecular Formula C51 H98 O6
Molecular Weight (g/mol) 807.32
Synonym Hexadecanoic acid, 1,1′,1′′-(1,2,3-propanetriyl) ester (ACI), Hexadecanoic acid, 1,2,3-propanetriyl ester (9CI), Palmitin, tri- (8CI), 2,3-Di(hexadecanoyloxy)propyl hexadecanoate, Barolub LCD, Dynasan 116, Dynosan 114, Glycerin tripalmitate, Glycerol tripalmitate, Glyceryl trihexadecanoate, Glyceryl tripalmitate, Palmitic acid triglyceride, Palmitic triglyceride, Spezialfett 116, Sterotex GTP, Triglyceride PPP, Triglyceryl palmitate, Tripalmitate, Tripalmitoylglycerol
IUPAC Name 2,3-di(hexadecanoyloxy)propyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C51H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-49(52)55-46-48(57-51(54)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)47-56-50(53)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h48H,4-47H2,1-3H3
Formula Weight 806.7363
Percent Purity >95%
Chemical Name or Material Tripalmitin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.