Learn More
Invitrogen™ TRIP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113945
Description
Immunogen sequence: MSGPTWLPPK QPEPARAPQG RAIPRGTPGP PPAHGAALQP HPRVNFCPLP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 86%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rat: 100%.
This gene is a member of the zyxin family and encodes a protein with three LIM zinc-binding domains. This protein localizes to focal adhesion sites and along actin stress fibers. Recruitment of this protein to the plasma membrane occurs in a lysophosphatidic acid (LPA)-dependent manner and it regulates LPA-induced cell migration. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
Specifications
| TRIP6 | |
| Polyclonal | |
| Unconjugated | |
| TRIP6 | |
| MGC10556; MGC10558; MGC29959; MGC3837; MGC4423; OIP1; OIP-1; OPA-interacting protein 1; tcag7.1300; thyroid hormone receptor interacting protein 6; thyroid hormone receptor interactor 6; thyroid receptor-interacting protein 6; TR-interacting protein 6; Trip6; TRIP-6; TRIP6i2; Zrp1; ZRP-1; zyxin related protein 1; zyxin-related protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7205 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q15654 | |
| TRIP6 | |
| Synthetic peptide directed towards the N terminal region of human TRIP6 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.