missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIP6 Antibody (6H4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37440-0.025ml
This item is not returnable.
View return policy
Description
TRIP6 Monoclonal specifically detects TRIP6 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| TRIP6 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| MGC10556, MGC10558, MGC29959, MGC3837, MGC4423, OIP1OIP-1, Opa-interacting protein 1, thyroid hormone receptor interactor 6, thyroid receptor-interacting protein 6, TR-interacting protein 6, ZRP-1TRIP-6, zyxin related protein 1, Zyxin-related protein 1 | |
| Mouse | |
| 50.3 kDa | |
| 0.025 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 7205 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 6H4 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| TRIP6 | |
| Purified recombinant fragment of human TRIP6 (AA: 107-291) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction