missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIP13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19602-1-AP-150UL
This item is not returnable.
View return policy
Description
Thyroid hormone receptors (TRs) are transcription factors that regulate the expression of specific genes in a hormone-dependent manner. TRIP13 (thyroid hormone receptor interactor 13), also called 16E1BP, is a transcription factor that interacts with the ligand binding domain of the thyroid receptor (TR) as well as a variety of target genes including human papilloma virus type 16 (HPV16) E1. Unlike most TRIP proteins which function only in the presence of hormones, TRIP13 does not require the presence of thyroid hormone to interact with TR. The association of TRIP13 with (HPV16) E1 suggests that TRIP13 may have tumor suppressor gene function. TRIP13 is a 432 amino acid protein with 2 different isoforms produced by alternative splicing.Specifications
| TRIP13 | |
| Polyclonal | |
| Unconjugated | |
| TRIP13 | |
| 16E1 BP, 16E1BP, PCH2, TR interacting protein 13, TRIP 13, TRIP13 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 292206, 69716, 9319 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15645, Q3UA06, Q5XHZ9 | |
| Trip13 | |
| TRIP13 Fusion Protein Ag6595 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction