missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIP13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531542
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431. Predicted reactivity: Mouse (95%), Rat (96%), Xenopus laevis (84%), Dog (94%), Pig (95%), Chicken (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Thyroid hormone receptors (TRs) are transcription factors that regulate the expression of specific genes in a hormone-dependent manner. TRIP13 (thyroid hormone receptor interactor 13), also called 16E1BP, is a transcription factor that interacts with the ligand binding domain of the thyroid receptor (TR) as well as a variety of target genes including human papilloma virus type 16 (HPV16) E1. Unlike most TRIP proteins which function only in the presence of hormones, TRIP13 does not require the presence of thyroid hormone to interact with TR. The association of TRIP13 with (HPV16) E1 suggests that TRIP13 may have tumor suppressor gene function. TRIP13 is a 432 amino acid protein with 2 different isoforms produced by alternative splicing.
Specifications
| TRIP13 | |
| Polyclonal | |
| Unconjugated | |
| TRIP13 | |
| 16E1BP; 16E1-BP; 2410002G23Rik; D13Ertd328e; HPV16 E1 protein binding protein; HPV16 E1 protein-binding protein; human papillomavirus type 16 E1 protein-binding protein; Pachytene checkpoint protein 2 homolog; PCH2; Thyroid hormone receptor interactor 13; thyroid receptor interacting protein 13; thyroid receptor-interacting protein 13; TR-interacting protein 13; TRIP13; TRIP-13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9319 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.88 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q15645 | |
| TRIP13 | |
| Recombinant protein encompassing a sequence within the center region of human TRIP13. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction