missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIP10 Recombinant Rabbit Monoclonal Antibody (23GB4705), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553791
This item is not returnable.
View return policy
Description
Required for translocation of GLUT4 to the plasma membrane in response to insulin signaling. Required to coordinate membrane tubulation with reorganization of the actin cytoskeleton during endocytosis. Binds to lipids such as phosphatidylinositol 4,5-bisphosphate and phosphatidylserine and promotes membrane invagination and the formation of tubules. Also promotes CDC42-induced actin polymerization by recruiting WASL/N-WASP which in turn activates the Arp2/3 complex. Actin polymerization may promote the fission of membrane tubules to form endocytic vesicles. Required for the formation of podosomes, actin-rich adhesion structures specific to monocyte-derived cells. May be required for the lysosomal retention of FASLG/FASL.
Specifications
| TRIP10 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Trip10 | |
| AI646975; Cdc42 interacting protein-4; cdc42-interacting protein 4; Cip4; hSTP; protein Felic; Salt tolerant protein; salt tolerator; STOT; STP; thyroid hormone receptor interactor 10; thyroid receptor-interacting protein 10; TR-interacting protein 10; TRIP10; TRIP-10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 106628 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4705 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8CJ53 | |
| Trip10 | |
| A synthesized peptide derived from human Cip4 (1-34AA). | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction