missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIP Monoclonal Antibody (OTI2D4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526366
This item is not returnable.
View return policy
Description
TRIP Monoclonal Antibody for Western Blot, IHC (P), Flow
TRAIP is a protein that contains an N-terminal RING finger motif and a putative coiled-coil domain. A similar murine protein interacts with TNFR-associated factor 1 (TRAF1), TNFR-associated factor 2 (TRAF2), and cylindromatosis. The interaction with TRAF2 inhibits TRAF2-mediated nuclear factor kappa-B, subunit 1 activation that is required for cell activation and protection against apoptosis.
Specifications
| TRIP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BWF2 | |
| TRAIP | |
| Human recombinant protein fragment corresponding to amino acids 67-313 of TRAIP produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D4 | |
| Unconjugated | |
| TRAIP | |
| E3 ubiquitin-protein ligase TRAIP; RING finger protein 206; RING-type E3 ubiquitin transferase TRAIP; RNF206; SCKL9; TRAF interacting protein; TRAF-interacting protein; TRAIP; Trip | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10293 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction