missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Triosephosphate isomerase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30012
This item is not returnable.
View return policy
Description
Triosephosphate isomerase Polyclonal antibody specifically detects Triosephosphate isomerase in Human,Mouse samples. It is validated for Western Blot
Specifications
| Triosephosphate isomerase | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| EC 5.3.1.1, MGC88108, TIM, TPI, triosephosphate isomerase, Triose-phosphate isomerase, triosephosphate isomerase 1 | |
| Between 199 and 249 | |
| 100 μg | |
| Stem Cell Markers | |
| 7167 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction