missing translation for 'onlineSavingsMsg'
Learn More

Trioctylmethylammonium Chloride, TRC

Product Code. 30457378
Change view
Click to view available options
Quantity:
25 mL
50 mL
100 mL
Unit Size:
100mL
25mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457378 25 mL 25mL
30449900 100 mL 100mL
30455306 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457378 Supplier Toronto Research Chemicals Supplier No. TRCT80445025ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyltrioctylammonium Chloride
CAS 5137-55-3
Molecular Formula C25 H54 N . Cl
Quantity 25 mL
Synonym Methyltrioctylammonium Chloride, A 336, A 336 (catalyst), Aliquat 128, Capriquat, Methyltri-n-octylammonium Chloride, Methyltricaprylammonium Chloride, N-Methyl-N,N,N-trioctylammonium Chloride, TOMAC, TOMAC 75, Tri-n-octylmethylammonium Chloride, Tricaprylmethylammonium Chloride, Trioctylmonomethylammonium Chloride
InChI Formula InChI=1S/C25H54N.ClH/c1-5-8-11-14-17-20-23-26(4,24-21-18-15-12-9-6-2)25-22-19-16-13-10-7-3;/h5-25H2,1-4H3;1H/q+1;/p-1
SMILES [Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC
IUPAC Name methyl(trioctyl)azanium;chloride
Molecular Weight (g/mol) 404.16
Formula Weight 403.3945
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.