missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TRIO Polyclonal antibody specifically detects TRIO in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TRIO |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ARHGEF23, EC 2.7.11.1, FLJ42780, PTPRF-interacting protein, tgat, triple functional domain (PTPRF interacting), triple functional domain protein |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 1575 to 1625 of human Trio Rho Guanine Nucleotide Exchange Factor using the numbering given in entry NP_009049.2 (GeneID 7204). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?