Learn More
Leica Microsystems Trinocular Tube
Product Code. 11974499
Change view
Click to view available options
For Use With (Equipment):
DM IL LED Inverted Microscope
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
11974499 DM IL LED Inverted Microscope Each
1 options
Product Code. 11974499 Supplier Leica Microsystems Supplier No. 11521249
This item requires a price update prior to being ordered.
Request a Price for this item here.
A new inverted microscope for routine and laboratory microscopy in cell biology and medicine
The Leica DM IL LED is a new inverted microscope for routine and laboratory microscopy in cell biology and medicine.
  • The perfect choice for applications in cell biology and medicine, micromanipulation, biotechnology, developmental biology, transgenics, molecular biology and fluorescence
  • Sturdy design with low stage height and plenty of space for operation
  • Long working distance objectives
  • Ergonomic arrangement of controls for fatigue free operation
  • Outstanding phase contrast for unstained samples
  • LED illumination provides low energy consumption and no heat build up
  • LEDs last 50,000 hours, eliminating the inconvenience of replacing lamps
TRUSTED_SUSTAINABILITY
Specifications
Type Trinocular Tube
For Use With (Application) Tube Program DM
For Use With (Equipment) DM IL LED Inverted Microscope
Quantity 1 Ea.