Learn More
Motic™ Trinocular Head, BA310 MET
Description
- Siedentopf type.
- Designed for rBA310 MET Microscopes.
- Inclined 30°.
Specifications
Specifications
| Type | Siedentopf, 30° Incline Trinocular Head |
| Description | Light Split: 100:0/50:50 |
| For Use With (Equipment) | BA310 MET Microscopes |