Learn More
Motic™ Trinocular Eyepiece Adapter, 28 Series
Product Code. 15415112
Change view
Click to view available options
For Use With (Equipment):
Series 28 Microscopes
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15415112 Series 28 Microscopes Each
1 options
Product Code. 15415112 Supplier Motic™ Supplier No. 1101000300592
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Facilitate photomicography with this adapter. Motic™ Trinocular Eyepiece Adapter, 28 Series allows you to connect a C-type SLR camera to your microscope.
  • Facilitates photomicography.
  • Designed for C-mount SLR cameras.
Specifications
Type C-type and SLR Camera Adapter
For Use With (Equipment) Series 28 Microscopes
Includes Eyepiece