missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimipramine Maleate, Mikromol™
Description
Trimipramine Maleate
Specifications
Specifications
| Chemical Name or Material | Trimipramine maleate |
| CAS | 521-78-8 |
| Analyte or Component Names | Trimipramine maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H26 N2 . C4 H4 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Trimipramine Maleate |
| InChI Formula | InChI=1S/C20H26N2.C4H4O4/c1-16(14-21(2)3)15-22-19-10-6-4-8-17(19)12-13-18-9-5-7-11-20(18)22;5-3(6)1-2-4(7)8/h4-11,16H,12-15H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CC(CN(C)C)CN1c2ccccc2CCc3ccccc13.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N,2-trimethylpropan-1-amine |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?