missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimipramine maleate, MedChemExpress
Description
Trimipramine maleate is also a potent and selective inhibitor targeting human noradrenaline (hNAT), serotonin (hSERT) and organic cation transporters (hOCT1, hOCT2) with IC50 values of 4.99 μM, 2.11 μM, 3.72 μM, 8.00 μM, respectively. Trimipramine maleate has vascular activity and anxiolytic efficacy.
Specifications
Specifications
| Chemical Name or Material | Trimipramine maleate |
| CAS | 521-78-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H30N2O4 |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (243.60 mM) ∣H2O : 14.29 mg/mL (34.81 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | CC(CN(C)C)CN1C2=C(C=CC=C2)CCC3=C1C=CC=C3.O=C(O)/C=C\C(O)=O |
| Molecular Weight (g/mol) | 410.51 |
| Formula Weight | 410.51 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?