missing translation for 'onlineSavingsMsg'
Learn More

Trimipramine.maleate, Lipomed

Product Code. 30461129
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461129 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461129 Supplier Lipomed Supplier No. LPMTRP1078MA100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Trimipramine.maleate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimipramine maleate
CAS 521-78-8
Analyte or Component Names Trimipramine maleate
Shipping Condition Room Temperature
Molecular Formula C20 H26 N2 . C4 H4 O4
Synonym Trimipramine Maleate
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CC(CN(C)C)CN1c2ccccc2CCc3ccccc13.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;3-(5,6-dihydrobenzo[b][1 ]benzazepin-11-yl)-N,N,2-trimethylpropan-1-amine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 410.51
Quantity 100 mg
Formula Weight 410.2206 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.