missing translation for 'onlineSavingsMsg'
Learn More

Trimetrexate, MedChemExpress

Product Code. 16476627
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476627

Brand: Medchem Express HY10373

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Trimetrexate (CI-898) is an antibiotic, also a potent and orally active dihydrofolate reductase (DHFR) inhibitor, reducing the production of DNA and RNA precursors and leading to cell death, with IC50 values of 4.74 nM and 1.35 nM for human DHFR and Toxoplasma gondii DHFR. Trimetrexate can also inhibit the growth of various cancer cells. Trimetrexate can be used for researching Pneumocystis carinii pneumonia (PCP) and cancer.

Trimetrexate (CI-898) is an antibiotic, also a potent and orally active dihydrofolate reductase (DHFR) inhibitor, reducing the production of DNA and RNA precursors and leading to cell death, with IC50 values of 4.74 nM and 1.35 nM for human DHFR and Toxoplasma gondii DHFR. Trimetrexate can also inhibit the growth of various cancer cells. Trimetrexate can be used for researching Pneumocystis carinii pneumonia (PCP) and cancer.

Specifications

Chemical Name or Material Trimetrexate
Physical Form Solid
CAS 52128-35-5
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H23N5O3
For Use With (Application) COVID-19-immunoregulation
Synonym CI-898
Solubility Information DMSO : ≥ 61.5 mg/mL (166.48 mM)
SMILES NC1=C(C(C)=C2CNC3=CC(OC)=C(OC)C(OC)=C3)C(C=C2)=NC(N)=N1
Molecular Weight (g/mol) 369.42
Formula Weight 369.42
Percent Purity 99.22%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.