missing translation for 'onlineSavingsMsg'
Learn More

Trimethylurea, TRC

Product Code. 30454934
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
Product Code. Quantity unitSize
30454934 500 mg 500mg
30452148 250 mg 250mg
30456257 2.5 g 2.5g
3 options
This item is not returnable. View return policy

Product Code. 30454934

Brand: Toronto Research Chemicals TRCT798188500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-14-4
Formula Weight 102.0793
InChI Formula InChI=1S/C4H10N2O/c1-5-4(7)6(2)3/h1-3H3,(H,5,7)
IUPAC Name 1,1,3-trimethylurea
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.14
SMILES CNC(=O)N(C)C
Synonym Trimethyl Urea
Chemical Name or Material Trimethylurea

Specifications

Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.