missing translation for 'onlineSavingsMsg'
Learn More

Trimethylurea

Product Code. 11387188
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387188 25 g 25g
11377188 1 g 1g
11367188 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387188 Supplier Thermo Scientific Alfa Aesar Supplier No. 016475.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 632-14-4 | C4H10N2O | 102.14 g/mol

It is used as pharmaceutical chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-14-4
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.14
MDL Number MFCD00014868
InChI Key COSWCAGTKRUTQV-UHFFFAOYSA-N
Synonym trimethylurea, n,n,n'-trimethylurea, urea, trimethyl, n,n'-dimethyl-n-methylurea, trimethylharnstoff, n,n',n'-trimethylurea, trimethylharnstoff german, urea, 1,1,3-trimethyl, unii-rmk4wz573b, urea, n,n,n'-trimethyl
PubChem CID 12435
IUPAC Name 1,1,3-trimethylurea
SMILES CNC(=O)N(C)C

Specifications

Melting Point 74°C to 75°C
Odor Characteristic
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 102.14
Physical Form Solid/Semi-solid
Chemical Name or Material Trimethylurea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.