missing translation for 'onlineSavingsMsg'
Learn More

Trimethyltin Hydroxide, TRC

Product Code. 30451744
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451744 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451744 Supplier Toronto Research Chemicals Supplier No. TRCT797615250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethyltin Hydroxide
CAS 56-24-6
Molecular Formula C3H10OSn
Quantity 250 mg
Synonym Hydroxytrimethylstannane, Hydroxytrimethyltin, Trimethylhydroxytin, Trimethylstannanol, Trimethylstannyl Hydroxide
InChI Formula InChI=1S/3CH3.H2O.Sn/h3*1H3;1H2;/q;;;;+1/p-1
SMILES C[Sn](C)(C)O
IUPAC Name hydroxy(trimethyl)stannane
Molecular Weight (g/mol) 180.82
Formula Weight 181.9754
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.