missing translation for 'onlineSavingsMsg'
Learn More

Trimethyltin hydroxide, 98%

Product Code. 11354859
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354859 25 g 25g
11364859 1 g 1g
11374859 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354859 Supplier Thermo Scientific Alfa Aesar Supplier No. 071167.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Trimethyltin hydroxide, 98%, C3H11OSn, CAS Number-56-24-6, hydroxytrimethyltin, trimethylstannyl hydroxide, trimethyl tin hydroxide, stannane, hydroxytrimethyl, tin, trimethyl-, hydroxide, trimethyltin hydroxide, hydroxytrimethylstannane, trimethylstannanol, trimethylhydroxytin, unii-10alt8dj2v, 25g | CAS: 56-24-6 | C3H10OSn | 180.82 g/mol

Trimethyltin hydroxide is used in the synthesis of modified thymines, which is useful as inhibitors of RNase A. It is also used in the preparation of ketoheterocycle inhibitors of fatty acid amide inhibitors. It reacts with 2,2-disubstituted benzothiazolines to form trialkyltin derivatives of sulphur containing Schiff base complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-24-6
Molecular Formula C3H10OSn
Molecular Weight (g/mol) 180.82
MDL Number MFCD00013929
InChI Key OJZNYUDKNVNEMV-UHFFFAOYSA-M
Synonym trimethyltin hydroxide, stannane, hydroxytrimethyl, trimethylstannanol, hydroxytrimethyltin, trimethyl tin hydroxide, trimethylhydroxytin, hydroxytrimethylstannane, trimethylstannyl hydroxide, unii-10alt8dj2v, tin, trimethyl-, hydroxide
PubChem CID 6328620
IUPAC Name trimethyltin;hydrate
SMILES C[Sn](C)(C)O

Specifications

Melting Point 114°C to 118°C
Color White
Odor Pungent
Linear Formula (CH3)3SnOH
Quantity 25 g
UN Number UN3146
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 180.8
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Trimethyltin hydroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.