missing translation for 'onlineSavingsMsg'
Learn More

Trimethyltin bromide, Thermo Scientific™

Product Code. 11364839
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364839 5 g 5g
11374839 1 g 1g
11384839 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11364839 Supplier Thermo Scientific Alfa Aesar Supplier No. 071153.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-44-0
Molecular Formula C3H11BrSn
Molecular Weight (g/mol) 245.74
MDL Number MFCD00000051
InChI Key KNPKGRQZYPSMBF-UHFFFAOYSA-N
Synonym trimethyltin bromide, bromotrimethyltin, stannane, bromotrimethyl, trimethylstannyl bromide, trimethyltin bromide 6ci, me3snbr, tin, bromotrimethyl, acmc-20ajcw, bromo trimethyl stannane, ch3 3snbr
PubChem CID 14015
IUPAC Name bromo(trimethyl)stannane
SMILES Br.C[SnH](C)C

Specifications

Melting Point 27°C
Boiling Point 165°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN2788
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 243.7
Physical Form Liquid
Chemical Name or Material Trimethyltin bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.