missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfoxonium chloride, 97%

Product Code. 10254574 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10254574 5 g Glass bottle 5g
10595134 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10254574 Supplier Thermo Scientific Acros Supplier No. 381520050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5034-06-0 | C3H9ClOS | 128.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5034-06-0
Molecular Formula C3H9ClOS
Molecular Weight (g/mol) 128.62
MDL Number MFCD00011898
InChI Key KQYWHJICYXXDSQ-UHFFFAOYSA-M
Synonym trimethylsulfoxonium chloride, trimethyloxosulfonium chloride, trimethylsulfonium chloride oxide, trimethyloxosulphonium chloride, sulfonium, trimethyl-, chloride, s-oxide, sulfoxonium, trimethyl-, chloride, sulfoxonium, trimethyl-, chloride 8ci,9ci, trimethyl oxo-??-sulfanylium chloride, c3h9os.cl, sulfoxonium, chloride
PubChem CID 197819
SMILES C[S+](=O)(C)C.[Cl-]

Specifications

Melting Point 226.0°C to 229.0°C
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)3S(O)Cl
Quantity 5 g
Formula Weight 128.62
Percent Purity 97%
Chemical Name or Material Trimethylsulfoxonium chloride
compliance-icons
Product Identifier
  • Trimethylsulfoxonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.