missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfonium bromide, 98%

Product Code. 11406127
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11406127

Brand: Thermo Scientific Alfa Aesar B25098.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3084-53-5 | C3H9BrS | 157.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3084-53-5
Molecular Formula C3H9BrS
Molecular Weight (g/mol) 157.069
MDL Number MFCD00051367
InChI Key GOTIICCWNAPLMN-UHFFFAOYSA-M
Synonym trimethylsulfonium bromide, trimethylsulfoniumbromide, sulfonium, trimethyl-, bromide, trimethylsulfanium bromide, unii-j59ncr7385, sulfonium, trimethyl-, bromide 1:1, acmc-209hj3, ksc492m3f
PubChem CID 2735154
IUPAC Name trimethylsulfanium;bromide
SMILES C[S+](C)C.[Br-]

Specifications

Melting Point 203°C (sublimation)
Quantity 500 g
Sensitivity Hygroscopic
Formula Weight 157.07
Percent Purity 98%
Chemical Name or Material Trimethylsulfonium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.