missing translation for 'onlineSavingsMsg'
Learn More

Trimethylstearylammonium Chloride, TRC

Product Code. 30450464
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450464 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450464 Supplier Toronto Research Chemicals Supplier No. TRCT8960031G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethyloctadecylammonium Chloride
CAS 112-03-8
Molecular Formula C21 H46 N . Cl
Quantity 1 g
Synonym 1-Octadecanaminium','N,N,N-trimethyl-','chloride (9CI), Ammonium','trimethyloctadecyl-','chloride (8CI), Trimethyloctadecylammonium chloride (6CI), AB 250, AB 250 (surfactant), Aliquat 7, Arcade T 800, Arquad 18, Arquad 18-50, Arquad 18-55, Arquad 18-63, Boraquat P 30, CA 2450, CA 2465, Catinal STC 25AQ, Catinal STC 25W, Catinal STC 70ET, Catinal STC 80, Catinal STS 80, Catiogen TMS, Cation AB, Cation AB 600, Cationic emulsifier 1831, Dehyquart B, Econol TMS 28, Electrostripper QE, FAC 1W, Genamin STAC, Genamin STACP, Kamin RMS, Lebon TM 18, Lipo Gard 18-63, Lipoquad T 800, Miconium STAC 65, Monostearyltrimethylammonium chloride, Nikkol CA 2450, Nikkol CA 2465, Nissan Cation AB, Nissan Cation AB 600, Octadecyltrimethylammonium chloride, Q-S-T 50, Quartamin 80W, Quartamin 86, Quartamin 869P, Quartamin 86P, Quartamin 86P Conc, Quartamin 86W, Quartamin P 85, Quaternium 10, Quaternium 1831, STAC, Stearimonium chloride, Steartrimonium chloride, Stearyltrimethylammonium chloride, Stearyltrimonium chloride, Trimethylstearylammonium chloride, Varisoft TSC
InChI Formula InChI=1S/C21H46N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(2,3)4;/h5-21H2,1-4H3;1H/q+1;/p-1
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)C
IUPAC Name trimethyl(octadecyl)azanium;chloride
Molecular Weight (g/mol) 348.05
Formula Weight 347.3319
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.