missing translation for 'onlineSavingsMsg'
Learn More

Trimethylstearylammonium Chloride, TRC

Código de producto. 30451112
Cambiar vista
Click to view available options
Quantity:
5 g
Tamaño de la unidad:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Tamaño de la unidad
30451112 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30451112 Proveedor Toronto Research Chemicals N.º de proveedor TRCT8960035G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethyloctadecylammonium Chloride
CAS 112-03-8
Molecular Formula C21 H46 N . Cl
Quantity 5 g
Synonym 1-Octadecanaminium','N,N,N-trimethyl-','chloride (9CI), Ammonium','trimethyloctadecyl-','chloride (8CI), Trimethyloctadecylammonium chloride (6CI), AB 250, AB 250 (surfactant), Aliquat 7, Arcade T 800, Arquad 18, Arquad 18-50, Arquad 18-55, Arquad 18-63, Boraquat P 30, CA 2450, CA 2465, Catinal STC 25AQ, Catinal STC 25W, Catinal STC 70ET, Catinal STC 80, Catinal STS 80, Catiogen TMS, Cation AB, Cation AB 600, Cationic emulsifier 1831, Dehyquart B, Econol TMS 28, Electrostripper QE, FAC 1W, Genamin STAC, Genamin STACP, Kamin RMS, Lebon TM 18, Lipo Gard 18-63, Lipoquad T 800, Miconium STAC 65, Monostearyltrimethylammonium chloride, Nikkol CA 2450, Nikkol CA 2465, Nissan Cation AB, Nissan Cation AB 600, Octadecyltrimethylammonium chloride, Q-S-T 50, Quartamin 80W, Quartamin 86, Quartamin 869P, Quartamin 86P, Quartamin 86P Conc, Quartamin 86W, Quartamin P 85, Quaternium 10, Quaternium 1831, STAC, Stearimonium chloride, Steartrimonium chloride, Stearyltrimethylammonium chloride, Stearyltrimonium chloride, Trimethylstearylammonium chloride, Varisoft TSC
InChI Formula InChI=1S/C21H46N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(2,3)4;/h5-21H2,1-4H3;1H/q+1;/p-1
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)C
IUPAC Name trimethyl(octadecyl)azanium;chloride
Molecular Weight (g/mol) 348.05
Formula Weight 347.3319
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.