missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl trifluoromethanesulfonate, 99%

Product Code. 11494204
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494204 10 g 10g
11404214 50 g 50g
11414214 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494204 Supplier Thermo Scientific Alfa Aesar Supplier No. A12535.09
  • ONLINE EXCLUSIVE PRICE
    £31.50
    £28.65 / 10g
    Save £2.85
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27607-77-8 | C4H9F3O3SSi | 222.253 g/mol

Trimethylsilyl trifluoromethanesulfonate is used to prepare powerful Lewis acid, difluoroboron triflate etherate in acetonitrile solvent. It is also used as a reagent in a Dieckmann-like cyclization of ester-imides and diesters. Further, it is used in the conversion of carbonyl compounds to their enol ethers. It is also employed for chemical glycosylation reactions. Its reactivity is similar to trimethylsilyl chloride and is also involved in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl trifluoromethanesulfonate is used to prepare powerful Lewis acid, difluoroboron triflate etherate in acetonitrile solvent. It is also used as a reagent in a Dieckmann-like cyclization of ester-imides and diesters. Further, it is used in the conversion of carbonyl compounds to their enol ethers. It is also employed for chemical glycosylation reactions. Its reactivity is similar to trimethylsilyl chloride and is also involved in organic synthesis.

Solubility
Miscible with aliphatic, aromatic hydrocarbons, haloalkanes and ethers. Immiscible with water.

Notes
Moisture and air sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27607-77-8
Molecular Formula C4H9F3O3SSi
Molecular Weight (g/mol) 222.253
MDL Number MFCD00000406
InChI Key FTVLMFQEYACZNP-UHFFFAOYSA-N
Synonym trimethylsilyl triflate, tmsotf, trifluoromethanesulfonic acid trimethylsilyl ester, trimethylsilyl trifluoromethylsulphonate, unii-z84v0cbh9j, methanesulfonic acid, trifluoro-, trimethylsilyl ester, trimethylsilyl trifluoromethylsulfonate, trimethylsllytrifluoromethanesulphonate, trimethylsilyl trifluoromethanesulphonate, trimethylsilyltrifluoromethanesulphonate
PubChem CID 65367
IUPAC Name trimethylsilyl trifluoromethanesulfonate
SMILES C[Si](C)(C)OS(=O)(=O)C(F)(F)F

Specifications

Density 1.225
Boiling Point 39°C to 40°C (12 mmHg)
Flash Point 25°C (77°F)
Linear Formula (CH3)3SiSO3CF3
Refractive Index 1.36
Quantity 10 g
UN Number UN2924
Beilstein 1868911
Sensitivity Moisture sensitive
Merck Index 14,9719
Solubility Information Miscible with aliphatic,aromatic hydrocarbons,haloalkanes and ethers. Immiscible with water.
Formula Weight 222.26
Percent Purity 99%
Chemical Name or Material Trimethylsilyl trifluoromethanesulfonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethylsilyl trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contain: Trimethylsilyl trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.