missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl trifluoromethanesulfonate, 99%

Product Code. 11404214
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11404214

Brand: Thermo Scientific Alfa Aesar A12535.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27607-77-8 | C4H9F3O3SSi | 222.253 g/mol

Trimethylsilyl trifluoromethanesulfonate is used to prepare powerful Lewis acid, difluoroboron triflate etherate in acetonitrile solvent. It is also used as a reagent in a Dieckmann-like cyclization of ester-imides and diesters. Further, it is used in the conversion of carbonyl compounds to their enol ethers. It is also employed for chemical glycosylation reactions. Its reactivity is similar to trimethylsilyl chloride and is also involved in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl trifluoromethanesulfonate is used to prepare powerful Lewis acid, difluoroboron triflate etherate in acetonitrile solvent. It is also used as a reagent in a Dieckmann-like cyclization of ester-imides and diesters. Further, it is used in the conversion of carbonyl compounds to their enol ethers. It is also employed for chemical glycosylation reactions. Its reactivity is similar to trimethylsilyl chloride and is also involved in organic synthesis.

Solubility
Miscible with aliphatic, aromatic hydrocarbons, haloalkanes and ethers. Immiscible with water.

Notes
Moisture and air sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27607-77-8
Molecular Formula C4H9F3O3SSi
Molecular Weight (g/mol) 222.253
MDL Number MFCD00000406
InChI Key FTVLMFQEYACZNP-UHFFFAOYSA-N
Synonym trimethylsilyl triflate, tmsotf, trifluoromethanesulfonic acid trimethylsilyl ester, trimethylsilyl trifluoromethylsulphonate, unii-z84v0cbh9j, methanesulfonic acid, trifluoro-, trimethylsilyl ester, trimethylsilyl trifluoromethylsulfonate, trimethylsllytrifluoromethanesulphonate, trimethylsilyl trifluoromethanesulphonate, trimethylsilyltrifluoromethanesulphonate
PubChem CID 65367
IUPAC Name trimethylsilyl trifluoromethanesulfonate
SMILES C[Si](C)(C)OS(=O)(=O)C(F)(F)F

Specifications

Density 1.225
Boiling Point 39°C to 40°C (12 mmHg)
Flash Point 25°C (77°F)
Linear Formula (CH3)3SiSO3CF3
Refractive Index 1.36
Quantity 50 g
UN Number UN2924
Beilstein 1868911
Sensitivity Moisture sensitive
Merck Index 14,9719
Solubility Information Miscible with aliphatic,aromatic hydrocarbons,haloalkanes and ethers. Immiscible with water.
Formula Weight 222.26
Percent Purity 99%
Chemical Name or Material Trimethylsilyl trifluoromethanesulfonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.