missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl trifluoromethanesulfonate, 99%

Product Code. 10042630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10042630 10 mL Glass bottle 10mL
10042640 50 mL Glass bottle 50mL
10237473 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10042630 Supplier Thermo Scientific Acros Supplier No. 209440100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27607-77-8 | C4H9F3O3SSi | 222.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27607-77-8
Molecular Formula C4H9F3O3SSi
Molecular Weight (g/mol) 222.26
MDL Number MFCD00000406
InChI Key FTVLMFQEYACZNP-UHFFFAOYSA-N
Synonym trimethylsilyl triflate, tmsotf, trifluoromethanesulfonic acid trimethylsilyl ester, trimethylsilyl trifluoromethylsulphonate, unii-z84v0cbh9j, methanesulfonic acid, trifluoro-, trimethylsilyl ester, trimethylsilyl trifluoromethylsulfonate, trimethylsllytrifluoromethanesulphonate, trimethylsilyl trifluoromethanesulphonate, trimethylsilyltrifluoromethanesulphonate
PubChem CID 65367
IUPAC Name trimethylsilyl trifluoromethanesulfonate
SMILES C[Si](C)(C)OS(=O)(=O)C(F)(F)F

Specifications

Color Colorless to Brown
Density 1.2250g/mL
Boiling Point 140°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CF3SO3Si(CH3)3
Refractive Index 1.3600 to 1.362
Quantity 10 mL
Fieser 08,514; 09,497; 10,438; 11,584; 12,543; 13,118; 14,333; 16,363; 17,379
Merck Index 15, 9891
Specific Gravity 1.225
Solubility Information Solubility in water: reacts
Formula Weight 222.26
Percent Purity 99%
Physical Form Fuming Liquid
Chemical Name or Material Trimethylsilyl trifluoromethanesulfonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethylsilyl trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.