missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl isothiocyanate, 94%

Product Code. 11322086
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322086 5 g 5g
11332086 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322086 Supplier Thermo Scientific Alfa Aesar Supplier No. L03277.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2290-65-5 | C4H9NSSi | 131.268 g/mol

Trimethylsilyl isothiocyanate is used as derivatizing reagent during the synthesis of amino acid thiohydantoins. It participates in ring opening reaction of N-substituted aziridines and cyclohexene oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl isothiocyanate is used as derivatizing reagent during the synthesis of amino acid thiohydantoins. It participates in ring opening reaction of N-substituted aziridines and cyclohexene oxide.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Incompatible materials are Strong oxidizing agentsAlcohols, Strong bases, Amines, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2290-65-5
Molecular Formula C4H9NSSi
Molecular Weight (g/mol) 131.268
MDL Number MFCD00004797
InChI Key XLTUPERVRFLGLJ-UHFFFAOYSA-N
Synonym trimethylsilyl isothiocyanate, silane, isothiocyanatotrimethyl, trimethylsilylisothiocyanate, isothiocyanato trimethyl silane, tmsisothiocyanate, tms-isothiocyanate, acmc-1clhj, trimethylsilyl-isothiocyanate, trimethylsilyl isothio-cyanate
PubChem CID 75297
IUPAC Name isothiocyanato(trimethyl)silane
SMILES C[Si](C)(C)N=C=S

Specifications

Melting Point -33°C
Density 0.932
Boiling Point 143°C to 144°C
Flash Point 29°C (84°F)
Odor Pungent
Refractive Index 1.482
Quantity 5 g
UN Number UN1992
Beilstein 1745262
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Reacts with water.
Formula Weight 131.27
Percent Purity 94%
Chemical Name or Material Trimethylsilyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.