missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Trimethylsilyl)diazomethane, approx. 2M solution in diethyl ether, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
Specifications
CAS Min % | 28.19, 67.36 |
CAS Max % | 32.64, 71.81 |
Color | White |
Density | 0.7700g/mL |
Flash Point | −35°C |
Packaging | Glass bottle |
Quantity | 100 mL |
Specific Gravity | 0.77 |
Formula Weight | 114.23 |
Physical Form | Crystalline Powder or Crystals |
Show More |
Safety and Handling
Product Identifier
- (Trimethylsilyl)diazomethane, ca. 2M solution in diethylether
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Flammable liquid Category 1
- Specific target organ toxicity Category 3
- H350-May cause cancer.
- H330-Fatal if inhaled.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H224-Extremely flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'