missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
5mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
Specifications
Specifications
| CAS Min % | 32.15, 64.47 |
| CAS Max % | 35.54, 67.85 |
| Color | Yellow |
| Density | 0.6750g/mL |
| Boiling Point | 96.0°C |
| Flash Point | −23°C |
| Packaging | Glass bottle |
| Linear Formula | (CH3)3SiCHN2 |
| Quantity | 500 mL |
| Specific Gravity | 0.675 |
| Show More |
Safety and Handling
Product Identifier
- (Trimethylsilyl)diazomethane (2.0M solution in hexanes)
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Carcinogenicity Category 1B
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H350-May cause cancer.
- H315-Causes skin irritation.
- H330-Fatal if inhaled.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- H370-Causes damage to organs.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P310-Immediately call a POISON CENTER/doctor/.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction