missing translation for 'onlineSavingsMsg'
Learn More

(Trimethylsilyl)diazomethane, 2M in hexanes

Product Code. 11342739
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342739 100 mL 100mL
11322739 5 mL 5mL
11332739 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342739 Supplier Thermo Scientific Alfa Aesar Supplier No. H26744.AE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18107-18-1 | C4H10N2Si | 114.223 g/mol

(Trimethylsilyl)diazomethane is used as a methylating agent in organic chemistry. It is utilized in Gas Chromatography- Mass-Spectrometry (GC-MS) for the analysis of various carboxylic compounds. It is involved as a reagent in the Doyle-Kirmse reaction with allyl sulfides and allyl amines. It is considered and used as a less-explosive replacement fordiazomethane. It is also used as a reactant for Unnatural alfa-amino acid derivatives containing gem-bisphosphonates through Michael addition reaction, imidazolo[2,1-b]benzothiazole derivatives and aigialomycin D analogues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Trimethylsilyl)diazomethane is used as a methylating agent in organic chemistry. It is utilized in Gas Chromatography- Mass-Spectrometry (GC-MS) for the analysis of various carboxylic compounds. It is involved as a reagent in the Doyle-Kirmse reaction with allyl sulfides and allyl amines. It is considered and used as a less-explosive replacement fordiazomethane. It is also used as a reactant for Unnatural alfa-amino acid derivatives containing gem-bisphosphonates through Michael addition reaction, imidazolo[2,1-b]benzothiazole derivatives and aigialomycin D analogues.

Solubility
Immiscible with water. Miscible with organic solvents.

Notes
(Trimethylsilyl)diazomethane (TMSD) in alcoholic solvents under acidic or basic conditions can lead to the formation of diazomethane. TMSD is under review by the National Toxicological Program for potential inhalation hazards.Incompatible with oxidizing agents, chlorine, fluorine and perchlorates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18107-18-1
Molecular Formula C4H10N2Si
Molecular Weight (g/mol) 114.223
MDL Number MFCD00053946
InChI Key ONDSBJMLAHVLMI-UHFFFAOYSA-N
Synonym trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4
PubChem CID 167693
IUPAC Name diazomethyl(trimethyl)silane
SMILES C[Si](C)(C)C=[N+]=[N-]

Specifications

Quantity 100 mL
UN Number UN3383
Beilstein 1902903
Flash Point −23°C (−10°F)
Solubility Information Immiscible with water. Miscible with organic solvents.
Formula Weight 114.22
Concentration or Composition (by Analyte or Components) 2M in hexanes
Sensitivity Light and moisture sensitive
Density 0.718
Chemical Name or Material (Trimethylsilyl)diazomethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.