missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl cyanide, 98%, AcroSeal™

Product Code. 11975871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11975871 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 11975871 Supplier Thermo Scientific Acros Supplier No. 422061000
Outlet Corner
Use promo code 28080
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 7677-24-9 | C4H9NSi | 99.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7677-24-9
Melting Point 11.0°C
Density 0.7440g/mL
Boiling Point 114.0°C to 117.0°C
Flash Point 1°C
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Molecular Formula C4H9NSi
Linear Formula (CH3)3SiCN
Refractive Index 1.3880 to 1.3920
MDL Number MFCD00001765
Quantity 100 mL
Fieser 04,542; 05,720; 06,632; 07,397; 08,133; 09,127; 10,112; 11,147; 12,148; 13,87; 14,107; 15,102; 17,89
Specific Gravity 0.744
Synonym trimethylsilyl cyanide, cyanotrimethylsilane, trimethylsilanecarbonitrile, trimethylsilylcyanide, silanecarbonitrile, trimethyl, trimethylsilylnitrile, tmscn, trimethyl silane cyanide, trimethylsilylcarbonitrile, tms cyanide
Solubility Information Solubility in water: reacts
InChI Key LEIMLDGFXIOXMT-UHFFFAOYSA-N
SMILES C[Si](C)(C)C#N
IUPAC Name trimethylsilylformonitrile
Molecular Weight (g/mol) 99.21
PubChem CID 82115
Formula Weight 99.21
Percent Purity 98%
Chemical Name or Material Trimethylsilyl cyanide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trimethylsilyl cyanide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 2
  • Acute toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.