missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl bromoacetate, 98+%

Product Code. 15438789
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438789 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15438789 Supplier Thermo Scientific Alfa Aesar Supplier No. L00961.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18291-80-0 | C5H11BrO2Si | 211.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 68°C to 69°C (17mmHg)
CAS 18291-80-0
Quantity 25 g
Flash Point 28°C (82°F)
Molecular Formula C5H11BrO2Si
MDL Number MFCD00000187
UN Number UN2920
Beilstein 1927560
Synonym trimethylsilyl bromoacetate, bromoacetic acid trimethylsilyl ester, acetic acid, bromo-, trimethylsilyl ester, acmc-209ek0, trimethylsilyl monobromoacetate, trimethylsilylbromoacetate, bromoacetic acid, tms derivative, zchhfmwudhxpfn-uhfffaoysa
InChI Key ZCHHFMWUDHXPFN-UHFFFAOYSA-N
SMILES C[Si](C)(C)OC(=O)CBr
IUPAC Name trimethylsilyl 2-bromoacetate
Molecular Weight (g/mol) 211.13
PubChem CID 87554
Formula Weight 211.13
Percent Purity ≥98%
Chemical Name or Material Trimethylsilyl bromoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.