missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl azide, 94%

Product Code. 11382685
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11382685

Brand: Thermo Scientific Alfa Aesar L00173.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4648-54-8 | C3H9N3Si | 115.21 g/mol

Trimethylsilyl azide is used as an azidonation reagent for amines, amides, aldehydes and ketones. It is involved in the preparation of alfa- and beta-siloxy azides from carbonyl compounds and epoxides. It is also used in the preparation of heterocyclic compounds and also acts as an effective substitute for hydrazoic acid. Further, it is used in the Oseltamivir total synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl azide is used as an azidonation reagent for amines, amides, aldehydes and ketones. It is involved in the preparation of alfa- and beta-siloxy azides from carbonyl compounds and epoxides. It is also used in the preparation of heterocyclic compounds and also acts as an effective substitute for hydrazoic acid. Further, it is used in the Oseltamivir total synthesis.

Solubility
Miscible with toluene, dichloromethane, diethyl ether and most organic solvents.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4648-54-8
Molecular Formula C3H9N3Si
Molecular Weight (g/mol) 115.21
MDL Number MFCD00001986
InChI Key SEDZOYHHAIAQIW-UHFFFAOYSA-N
Synonym trimethylsilyl azide, silane, azidotrimethyl, trimethylsilylazide, azido trimethyl silane, ccris 8048, tms azide, tms-azide, trimethyl silylazide, trimethylsilyl-azide, azido trimethylsilane
PubChem CID 78378
SMILES C[Si](C)(C)N=[N+]=[N-]

Specifications

Melting Point -95°C
Density 0.872
Boiling Point 95°C to 99°C
Flash Point 6°C (42°F)
Refractive Index 1.414
Quantity 100 g
UN Number UN1992
Beilstein 1903730
Sensitivity Moisture sensitive
Solubility Information Miscible with toluene,dichloromethane,diethyl ether and most organic solvents.
IUPAC Name azido(trimethyl)silane
Formula Weight 115.21
Percent Purity 94%
Chemical Name or Material Trimethylsilyl azide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.