missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl acetate, 97%

Product Code. 11449527
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449527 100 g 100g
11439527 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449527 Supplier Thermo Scientific Alfa Aesar Supplier No. B22037.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2754-27-0 | C5H12O2Si | 132.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2754-27-0
Molecular Formula C5H12O2Si
Molecular Weight (g/mol) 132.234
MDL Number MFCD00008695
InChI Key QHUNJMXHQHHWQP-UHFFFAOYSA-N
Synonym acetoxytrimethylsilane, trimethylacetoxysilane, acetato trimethylsilane, acetic acid trimethylsilyl ester, silanol, trimethyl-, acetate, silanol, 1,1,1-trimethyl-, 1-acetate, tmsoac, o-trimethylsilylacetate, trimethylsilyl acetate
PubChem CID 75988
IUPAC Name trimethylsilyl acetate
SMILES CC(=O)O[Si](C)(C)C

Specifications

Melting Point -32°C
Density 0.884
Boiling Point 107°C
Flash Point 4°C (39°F)
Linear Formula CH3CO2Si(CH3)3
Refractive Index 1.388
Quantity 100 g
UN Number UN3272
Beilstein 1744463
Sensitivity Moisture sensitive
Formula Weight 132.24
Percent Purity 97%
Chemical Name or Material Trimethylsilyl acetate
Show More Show Less
compliance-icons
Product Identifier
  • Trimethylsilyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contain: Trimethylsilyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.