missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 94%

Product Code. 11357029
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357029 25 g 25g
15447625 1 g 1g
11347029 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357029 Supplier Thermo Scientific Alfa Aesar Supplier No. H26857.14
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 120801-75-4 | C5H9F3O4SSi | 250.26 g/mol

Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate is an fluoroalkylation agent, reactant for preparation of difluorocyclopropenes, preparation of difluorocarbene reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate is an fluoroalkylation agent, reactant for preparation of difluorocyclopropenes, preparation of difluorocarbene reagents.

Solubility
Reacts with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120801-75-4
Molecular Formula C5H9F3O4SSi
Molecular Weight (g/mol) 250.26
MDL Number MFCD02093343
InChI Key XHVSCKNABCCCAC-UHFFFAOYSA-N
PubChem CID 2778022
IUPAC Name trimethylsilyl 2,2-difluoro-2-fluorosulfonylacetate
SMILES C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O

Specifications

Density 1.27
Boiling Point 156°C
Flash Point 26°C (79°F)
Refractive Index 1.367
Quantity 25 g
UN Number UN2920
Synonym trimethylsilyl 2,2-difluoro-2-fluorosulfonyl acetate, trimethylsilyl 2-fluorosulfonyl difluoroacetate, trimethylsilyl2-fluorosulphonyl difluoroacetate, trimethylsilyl fluorosulfonyldifluoroacetate, trimethylsilyl difluoro fluorosulfonyl acetate, trimethylsilyl 2-fluorosulphonyl difluoroacetate, 2,2-difluoro-2-fluorosulfonyl acetic acid trimethylsilyl ester, pubchem22642, g00021-watson-int, acmc-1c7yr
Solubility Information Reacts with water.
Formula Weight 250.27
Percent Purity 94%
Chemical Name or Material Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.