missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilanol

Product Code. 15545286 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15545286 5 g 5g
15555286 25 g 25g
15565286 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15545286 Supplier Thermo Scientific Acros Supplier No. 458570050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1066-40-6 | C3H9LiOSi | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-40-6
Molecular Formula C3H9LiOSi
Molecular Weight (g/mol) 96.13
MDL Number MFCD02751657
InChI Key OXOZHAWWRPCVGL-UHFFFAOYSA-N
Synonym trimethylsilanol, silanol, trimethyl, unii-z4bin3300p, trimethylhydroxysilane, ccris 1320, hydroxy trimethyl silane, silanol, 1,1,1-trimethyl, ksc504m9b, 2004-14-0 lithium salt, 10519-96-7 potassium salt
PubChem CID 66110
IUPAC Name hydroxy(trimethyl)silane
SMILES [Li+].C[Si](C)(C)[O-]

Specifications

Melting Point -12.0°C
Density 0.8140g/mL
Boiling Point 98.0°C
Flash Point 4°C
Infrared Spectrum Authentic
Linear Formula (CH3)3SiOH
Quantity 5 g
Specific Gravity 0.814
Solubility Information Solubility in water: 0.995g/L.
Viscosity 4 mPas (25°C)
Formula Weight 90.2
Percent Purity ≥97.5% (GC)
Chemical Name or Material Trimethylsilanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethylsilanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.