missing translation for 'onlineSavingsMsg'
Learn More

Trimethylpyruvic acid, ca. 60% aq. soln.

Product Code. 11497256
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497256 250 g 250g
11487256 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497256 Supplier Thermo Scientific Alfa Aesar Supplier No. B25582.30
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 815-17-8 | C6H10O3 | 130.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 815-17-8
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00154352
InChI Key IAWVHZJZHDSEOC-UHFFFAOYSA-N
Synonym 3,3-dimethyl-2-oxobutyric acid, trimethylpyruvic acid, butanoic acid, 3,3-dimethyl-2-oxo, pyruvic acid, trimethyl, glyoxylic acid, tert-butyl, unii-29cl615clh, 3,3-dimethyl-2-oxo-butanoic acid, butyric acid, 3,3-dimethyl-2-oxo, tert-butylglyoxylic acid, trimethlpyruvic acid
PubChem CID 13150
IUPAC Name 3,3-dimethyl-2-oxobutanoic acid
SMILES CC(C)(C)C(=O)C(=O)O

Specifications

Linear Formula (CH3)3CCOCO2H
Quantity 250 g
UN Number UN3265
Sensitivity May form a precipitate
Formula Weight 130.14
Concentration or Composition (by Analyte or Components) ≈60% aq. Soln.
Chemical Name or Material Trimethylpyruvic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.