missing translation for 'onlineSavingsMsg'
Learn More

Trimethylphosphine oxide

Product Code. 11357268
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357268 1 g 1g
11367268 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357268 Supplier Thermo Scientific Alfa Aesar Supplier No. 030394.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 676-96-0 | C3H9OP | 92.078 g/mol

Trimethylphosphine oxide is a common side product in organic synthesis, including the Wittig, Staudinger, Mitsunobu reactions and also in the manufacture of triphenylphosphine. It is a popular reagent to induce the crystallization of various chemical compounds. It acts as an excellent ligand for hard metal centers. It can react with 9-methyl-9-azonianthracene methyl sulfate to get (10-methyl-9,10-dihydro-acridin-9-yl)-phosphonic acid dimethyl ester using sodium iodide and acetonitrile as solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 676-96-0
Molecular Formula C3H9OP
Molecular Weight (g/mol) 92.078
MDL Number MFCD00013910
InChI Key LRMLWYXJORUTBG-UHFFFAOYSA-N
Synonym trimethylphosphine oxide, phosphine oxide, trimethyl, ch3 3po, phosphine oxide,trimethyl-6ci,8ci,9ci, dimethylphosphoryl methane, trimethylphosphane oxide, trimethylphosphineoxide, me3po, acmc-1bhzf, trimethyloxylatophosphonium
PubChem CID 69609
IUPAC Name dimethylphosphorylmethane
SMILES CP(=O)(C)C

Specifications

Melting Point 140°C to 141°C
Odor Odorless
Quantity 1 g
Solubility Information Soluble in polar organic solvents.
Formula Weight 92.08
Physical Form Crystalline
Chemical Name or Material Trimethylphosphine oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.