missing translation for 'onlineSavingsMsg'
Learn More

Trimethylphosphine, 98%

Product Code. 15431167
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15431167 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15431167 Supplier Thermo Scientific Alfa Aesar Supplier No. 030143.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 594-09-2 | C3H9P | 76.079 g/mol

It is used in the preparation of organic phosphorus compounds and frequently-used phosphine ligand. It is also a highly basic ligand that forms complexes with most metals. In addition, it can react with bromomethyl-benzene to get benzyl-trimethyl-phosphonium; bromide. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-09-2
Molecular Formula C3H9P
Molecular Weight (g/mol) 76.079
MDL Number MFCD00008510
InChI Key YWWDBCBWQNCYNR-UHFFFAOYSA-N
Synonym trimethylphosphine, trimethyl phosphine, pme3, trimethylphosphorus, phosphine, trimethyl, me3p, ch3 3p, trimethylphospine, trimethylphosphine?, phosphine,trimethyl
PubChem CID 68983
ChEBI CHEBI:35890
IUPAC Name trimethylphosphane
SMILES CP(C)C

Specifications

Density 0.735 g/mL
Boiling Point 38°C to 40°C
Flash Point -30°C (-22°F)
Odor Pungent
Quantity 25 g
UN Number UN2845
Sensitivity Air and moisture sensitive
Solubility Information Soluble in water.
Formula Weight 76.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Trimethylphosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.