missing translation for 'onlineSavingsMsg'
Learn More

Trimethylphloroglucinol, TRC

Product Code. 30137860
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137860 5 mg 5mg
30135822 10 mg 10mg
30004266 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137860 Supplier Toronto Research Chemicals Supplier No. TRCT7966005MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4463-03-0
Molecular Formula C9 H12 O3
Molecular Weight (g/mol) 168.19
Synonym 2,4,6-trimethylbenzene-1,3,5-triol, 2,4,6-Trimethyl-1,3,5-benzenetriol;
IUPAC Name 2,4,6-trimethylbenzene-1,3,5-triol
SMILES Cc1c(O)c(C)c(O)c(C)c1O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C9H12O3/c1-4-7(10)5(2)9(12)6(3)8(4)11/h10-12H,1-3H3
Formula Weight 168.079
Chemical Name or Material Trimethylphloroglucinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.