Learn More
Trimethylolpropane triacrylate, stabilized, Thermo Scientific Chemicals
£35.20 - £220.00
Chemical Identifiers
CAS | 15625-89-5 |
---|---|
Molecular Formula | C15H20O6 |
Molecular Weight (g/mol) | 296.32 |
MDL Number | MFCD00008628 |
InChI Key | DAKWPKUUDNSNPN-UHFFFAOYSA-N |
Synonym | trimethylolpropane triacrylate, tmpta, viscoat 295, nk ester a tmpt, sartomer sr 351, ogumont t 200, saret 351, setalux uv 2241, ccris 92, unii-4b67kgl96s |
PubChem CID | 27423 |
ChEBI | CHEBI:35028 |
IUPAC Name | 2,2-bis(prop-2-enoyloxymethyl)butyl prop-2-enoate |
SMILES | CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337188
|
Thermo Scientific Alfa Aesar
016445.22 |
100 g |
£35.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357188
|
Thermo Scientific Alfa Aesar
016445.36 |
500 g |
£75.70
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347188
|
Thermo Scientific Alfa Aesar
016445.A3 |
2 kg |
£220.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15625-89-5 | |
296.32 | |
DAKWPKUUDNSNPN-UHFFFAOYSA-N | |
27423 | |
2,2-bis(prop-2-enoyloxymethyl)butyl prop-2-enoate |
C15H20O6 | |
MFCD00008628 | |
trimethylolpropane triacrylate, tmpta, viscoat 295, nk ester a tmpt, sartomer sr 351, ogumont t 200, saret 351, setalux uv 2241, ccris 92, unii-4b67kgl96s | |
CHEBI:35028 | |
CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C |
Specifications
15625-89-5 | |
>200°C (1 mmHg) | |
Pungent | |
1.4735 | |
trimethylolpropane triacrylate, tmpta, viscoat 295, nk ester a tmpt, sartomer sr 351, ogumont t 200, saret 351, setalux uv 2241, ccris 92, unii-4b67kgl96s | |
DAKWPKUUDNSNPN-UHFFFAOYSA-N | |
2,2-bis(prop-2-enoyloxymethyl)butyl prop-2-enoate | |
27423 | |
296.31 | |
Trimethylolpropane triacrylate, Stabilized |
1.10 g/mL | |
87°C (188°F) | |
C15H20O6 | |
MFCD00008628 | |
Miscible with water. | |
CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C | |
296.32 | |
CHEBI:35028 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319-H317-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Combustible liquid.
P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P370+P378q-P501c
H227-H315-H317-H319
missing translation for 'einecsNumber' : 239-701-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Trimethylolpropane triacrylate
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Trimethylolpropane triacrylate
RUO – Research Use Only