missing translation for 'onlineSavingsMsg'
Learn More

Trimethylolpropane triacrylate, stabilized

Product Code. 11347188
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347188 2 kg 2kg
11337188 100 g 100g
11357188 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11347188 Supplier Thermo Scientific Alfa Aesar Supplier No. 016445.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15625-89-5 | C15H20O6 | 296.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15625-89-5
Molecular Formula C15H20O6
Molecular Weight (g/mol) 296.32
MDL Number MFCD00008628
InChI Key DAKWPKUUDNSNPN-UHFFFAOYSA-N
Synonym trimethylolpropane triacrylate, tmpta, viscoat 295, nk ester a tmpt, sartomer sr 351, ogumont t 200, saret 351, setalux uv 2241, ccris 92, unii-4b67kgl96s
PubChem CID 27423
ChEBI CHEBI:35028
IUPAC Name 2,2-bis(prop-2-enoyloxymethyl)butyl prop-2-enoate
SMILES CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C

Specifications

Density 1.10 g/mL
Boiling Point >200°C (1 mmHg)
Flash Point 87°C (188°F)
Odor Pungent
Refractive Index 1.4735
Quantity 2 kg
Solubility Information Miscible with water.
Formula Weight 296.31
Physical Form Liquid
Chemical Name or Material Trimethylolpropane triacrylate, Stabilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.